Learn More
Biomatik Corporation Mouse Syndecan 1 (SDC1) ELISA Kit
Supplier:  Biomatik Corporation EKU07549
Encompass_Preferred
Mouse Syndecan 1 (SDC1) ELISA Kit for detecting SDC1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18828. Gene ID: 20969. Target Synonyms: CD138; SDC; SYND1; Syndecan Proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6400