Learn More
Biomatik Corporation Mouse Thrombin/Antithrombin Complex (TAT) ELISA Kit
Supplier:  Biomatik Corporation EKU07626
Encompass_Preferred
Mouse Thrombin/Antithrombin Complex (TAT) ELISA Kit for detecting TAT in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19221; P32261. Gene ID: 14061; 11905. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6408