Learn More
Biomatik Corporation Mouse Thymic Stromal Lymphopoietin (TSLP) ELISA Kit
Supplier:  Biomatik Corporation EKU07657
Encompass_Preferred
Mouse Thymic Stromal Lymphopoietin (TSLP) ELISA Kit for detecting TSLP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JIE6. Gene ID: 53603. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6412