Learn More
Biomatik Corporation Rat Vascular Endothelial Growth Factor C (VEGFC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08099
Encompass_Preferred
Rat Vascular Endothelial Growth Factor C (VEGFC) ELISA Kit for detecting VEGFC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35757. Gene ID: 114111. Target Synonyms: VEGF-C; Flt4-L; VRPL; Vascular Endothelial Growth Factor-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4109