Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit, 96 tests
Catalog No. 501504111
Encompass_Preferred
Human Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit for detecting VEGFR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17948. Gene ID: 2321. Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4111 Supplier Biomatik Corporation Supplier No. EKU08102
Human Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit for detecting VEGFR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17948. Gene ID: 2321. Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.