Learn More
Biomatik Corporation Rat Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08105
Encompass_Preferred
Rat Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit for detecting VEGFR1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53767. Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4113