Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08106
Encompass_Preferred
Human Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit for detecting VEGFR2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.236ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35968. Gene ID: 3791. Target Synonyms: CD309; FLK1; VEGFR; KDR; A Type III Receptor Tyrosine Kinase; Kinase Insert Domain Receptor; Kinase Insert Domain Receptor; Fetal Liver Kinase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4114