Learn More
Biomatik Corporation Human Ubiquitin Specific Peptidase 8 (USP8) ELISA Kit
Supplier:  Biomatik Corporation EKU08026
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.125ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P40818. Target Synonyms: HumORF8; UBPY; Ubiquitin Carboxyl-Terminal Hydrolase 8; Ubiquitin Specific Protease 8; Deubiquitinating enzyme 8; Ubiquitin isopeptidase Y; Ubiquitin thioesterase 8. Restriction: For Research Use Only.
Catalog No. 50-228-6455