Learn More
Biomatik Corporation Rat Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08109
Encompass_Preferred
Rat Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit for detecting VEGFR2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 24.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08775. Gene ID: 25589. Target Synonyms: CD309; FLK1; VEGFR; KDR; A Type III Receptor Tyrosine Kinase; Kinase Insert Domain Receptor; Kinase Insert Domain Receptor; Fetal Liver Kinase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4116