Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor Receptor 3 (VEGFR3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08110
Encompass_Preferred
Human Vascular Endothelial Growth Factor Receptor 3 (VEGFR3) ELISA Kit for detecting VEGFR3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35916. Gene ID: 2324. Target Synonyms: FLT41; FLT4; PCL; Fms-Related Tyrosine Kinase 4; Tyrosine-protein kinase receptor FLT4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4117