Learn More
Biomatik Corporation Rat Vascular Endothelial Growth Factor Receptor 3 (VEGFR3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08111
Encompass_Preferred
Rat Vascular Endothelial Growth Factor Receptor 3 (VEGFR3) ELISA Kit for detecting VEGFR3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91ZT1. Gene ID: 114110. Target Synonyms: FLT41; FLT4; PCL; Fms-Related Tyrosine Kinase 4; Tyrosine-protein kinase receptor FLT4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4118