Learn More
Biomatik Corporation Human Vasoactive Intestinal Peptide (VIP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08113
Encompass_Preferred
Human Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.63pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01282. Gene ID: 7432. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4119