Learn More
Biomatik Corporation Mouse Vasoactive Intestinal Peptide (VIP) ELISA Kit, 96 tests
Catalog No. 501504120
Encompass_Preferred
Mouse Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.71pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P32648. Gene ID: 22353. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4120 Supplier Biomatik Corporation Supplier No. EKU08114
Mouse Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.71pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P32648. Gene ID: 22353. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.