Learn More
Biomatik Corporation Rat Vasoactive Intestinal Peptide (VIP) ELISA Kit, 96 tests
Catalog No. 501504121
Encompass_Preferred
Rat Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.58pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01283. Gene ID: 117064. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4121 Supplier Biomatik Corporation Supplier No. EKU08115
Rat Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.58pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01283. Gene ID: 117064. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.