Learn More
Biomatik Corporation Human Interleukin 2 (IL2) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU08274
Encompass_Preferred
Human Interleukin 2 (IL2) High-sensitive ELISA Kit for detecting IL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60568. Gene ID: 3558. Target Synonyms: TCGF; Lymphokine; T-Cell Growth Factor; Aldesleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6485