Learn More
Biomatik Corporation Mouse VEGF Co Regulated Chemokine 1 (VCC1) ELISA Kit, 96 tests
Catalog No. 501504124
Encompass_Preferred
Mouse VEGF Co Regulated Chemokine 1 (VCC1) ELISA Kit for detecting VCC1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5UW37. Gene ID: 232983. Target Synonyms: CXCL17; SCYB17; DMC; Chemokine C-X-C-Motif Ligand 7; Dendritic cell and monocyte chemokine-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4124 Supplier Biomatik Corporation Supplier No. EKU08122
Mouse VEGF Co Regulated Chemokine 1 (VCC1) ELISA Kit for detecting VCC1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5UW37. Gene ID: 232983. Target Synonyms: CXCL17; SCYB17; DMC; Chemokine C-X-C-Motif Ligand 7; Dendritic cell and monocyte chemokine-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.