Learn More
Biomatik Corporation Rabbit Collagen Type II (COL2) ELISA Kit
Supplier:  Biomatik Corporation EKU08459
Encompass_Preferred
Rabbit Collagen Type II (COL2) ELISA Kit for detecting COL2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1T5V9. Gene ID: 100009005. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6501