Learn More
Biomatik Corporation Mouse Versican (VS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08128
Encompass_Preferred
Mouse Versican (VCAN) ELISA Kit for detecting VCAN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62059. Target Synonyms: CSPG2; VS; ERVR; PG-M; WGN1; Chondroitin Sulfate Proteoglycan 2; Chondroitin sulfate proteoglycan core protein 2; Glial hyaluronate-binding; Large fibroblast proteoglycan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4127