Learn More
Biomatik Corporation Human VGF Nerve Growth Factor Inducible (VGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08139
Encompass_Preferred
Human VGF Nerve Growth Factor Inducible (VGF) ELISA Kit for detecting VGF in serum, plasma, cerebrospinal fluid, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15240. Gene ID: 7425. Target Synonyms: NERP1; NERP2; Neuro-Endocrine Specific Protein VGF; Neuroendocrine Regulatory Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4128