Learn More
Biomatik Corporation Mouse VGF Nerve Growth Factor Inducible (VGF) ELISA Kit, 96 tests
Catalog No. 501504129
Encompass_Preferred
Mouse VGF Nerve Growth Factor Inducible (VGF) ELISA Kit for detecting VGF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q0VGU4. Gene ID: 381677. Target Synonyms: NERP1; NERP2; Neuro-Endocrine Specific Protein VGF; Neuroendocrine Regulatory Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4129 Supplier Biomatik Corporation Supplier No. EKU08140
Mouse VGF Nerve Growth Factor Inducible (VGF) ELISA Kit for detecting VGF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q0VGU4. Gene ID: 381677. Target Synonyms: NERP1; NERP2; Neuro-Endocrine Specific Protein VGF; Neuroendocrine Regulatory Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.