Learn More
Biomatik Corporation Human Virus Inhibitory Protein (Viperin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08147
Encompass_Preferred
Human Virus Inhibitory Protein (Viperin) ELISA Kit for detecting Viperin in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8WXG1. Gene ID: 91543. Target Synonyms: Cig5; RSAD2; vig1; Radical S-Adenosyl Methionine Domain Containing 2; Viperin,Endoplasmic Reticulum-Associated,Interferon-Inducible; Cytomegalovirus-induced gene 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4132