Learn More
Biomatik Corporation Mouse Visceral Adipose Tissue Derived Serine Protease Inhibitor (Vaspin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08150
Encompass_Preferred
Mouse Visceral Adipose Tissue Derived Serine Protease Inhibitor (Vaspin) ELISA Kit for detecting Vaspin in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TMF5. Gene ID: 68054. Target Synonyms: SERPINA12; OL-64; Visceral Adipose-Specific SERPIN; Serine Proteinase Inhibitor,Clade A Member 12; Visceral adipose-specific serpin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4134