Learn More
Biomatik Corporation Vitamin K1 (VK1) ELISA Kit
Supplier:  Biomatik Corporation EKU08716
Encompass_Preferred
Vitamin K1 (VK1) ELISA Kit for detecting VK1 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000pg/ml. Sensitivity: 55.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Phylloquinone; Phytonadione; Phytomenadione. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6531