Learn More
Biomatik Corporation Human Advanced Oxidation Protein Products (AOPP) ELISA Kit
Supplier:  Biomatik Corporation EKU08721
Encompass_Preferred
Human Advanced Oxidation Protein Products (AOPP) ELISA Kit for detecting AOPP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 61.7-5,000ng/ml. Sensitivity: 25.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6533