Learn More
Biomatik Corporation Diacylglycerol (DAG) ELISA Kit
Supplier:  Biomatik Corporation EKU08737
Encompass_Preferred
Diacylglycerol (DAG) ELISA Kit for detecting DAG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 391-100,000pg/ml. Sensitivity: 153pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: DG; Diglyceride. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6538