Learn More
Biomatik Corporation Human Visinin Like Protein 1 (VSNL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08157
Encompass_Preferred
Human Visinin Like Protein 1 (VSNL1) ELISA Kit for detecting VSNL1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62760. Gene ID: 7447. Target Synonyms: HLP3; VISL1; HPCAL3; HUVISL1; VILIP; VILIP-1; VLP-1; Hippocalcin Like Protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4138