Learn More
Biomatik Corporation Human Vitamin D Binding Protein (DBP) ELISA Kit, 96 tests
Catalog No. 501504140
Encompass_Preferred
Human Vitamin D Binding Protein (DBP) ELISA Kit for detecting DBP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.072ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02774. Gene ID: 2638. Target Synonyms: GC; VDBG; VDBP; Group-Specific Component; Gc-globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4140 Supplier Biomatik Corporation Supplier No. EKU08164
Human Vitamin D Binding Protein (DBP) ELISA Kit for detecting DBP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.072ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02774. Gene ID: 2638. Target Synonyms: GC; VDBG; VDBP; Group-Specific Component; Gc-globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.