Learn More
Biomatik Corporation Taurocholic Acid (TCA) ELISA Kit
Supplier:  Biomatik Corporation EKU08759
Encompass_Preferred
Taurocholic Acid (TCA) ELISA Kit for detecting TCA in biological fluids. Species Reactivity: Universal. Detection Range: 3.70-300ng/ml. Sensitivity: 1.33ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Cholaic Acid; Cholyltaurine; Acidum Cholatauricum. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6545