Learn More
Biomatik Corporation Mouse Vitamin D Receptor (VDR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08168
Encompass_Preferred
Mouse Vitamin D Receptor (VDR) ELISA Kit for detecting VDR in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48281. Gene ID: 22337. Target Synonyms: NR1I1; Nuclear Receptor Subfamily 1 Group I Member 1; Calcitriol Receptor; 1,25-dihydroxyvitamin D3 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4142