Learn More
Biomatik Corporation Butyric Acid (BA) ELISA Kit
Supplier:  Biomatik Corporation EKU08762
Encompass_Preferred
Butyric Acid (BA) ELISA Kit for detecting BA in biological agents. Species Reactivity: Universal. Detection Range: 123.5-10,000pg/ml. Sensitivity: 56.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Propanecarboxylic Acid; Butanoic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6547