Learn More
Biomatik Corporation Mouse Vitronectin (VTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08173
Encompass_Preferred
Mouse Vitronectin (VTN) ELISA Kit for detecting VTN in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29788. Gene ID: 22370. Target Synonyms: V75; VN; VNT; Serum Spreading Factor; Somatomedin B; Complement S-Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4144