Learn More
Biomatik Corporation Human V-Ki-Ras2 Kirsten Rat Sarcoma Viral Oncogene Homolog (KRAS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08176
Encompass_Preferred
Human V-Ki-Ras2 Kirsten Rat Sarcoma Viral Oncogene Homolog (KRAS) ELISA Kit for detecting KRAS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01116. Gene ID: 3845. Target Synonyms: C-K-RAS; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS1; KRAS2; NS3; RASK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4145