Learn More
Biomatik Corporation Human Voltage Dependent Anion Channel Protein 1 (VDAC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08181
Encompass_Preferred
Human Voltage Dependent Anion Channel Protein 1 (VDAC1) ELISA Kit for detecting VDAC1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21796. Target Synonyms: PORIN-31-HL; Porin 31HM; Plasmalemmal porin; Outer mitochondrial membrane protein porin 1; Voltage-dependent anion-selective channel protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4146