Learn More
Biomatik Corporation Rabbit Von Willebrand Factor (vWF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08188
Encompass_Preferred
Rabbit Von Willebrand Factor (vWF) ELISA Kit for detecting vWF in plasma. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 4.64ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q28834. Target Synonyms: F8VWF; VWD; von Willebrand antigen 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4148