Learn More
Biomatik Corporation Human V-Rel Reticuloendotheliosis Viral Oncogene Homolog A (RELA) ELISA Kit, 96 tests
Catalog No. 501504149
Encompass_Preferred
Human Transcription Factor P65 (RELA) ELISA Kit for detecting RELA in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04206. Gene ID: 5970. Target Synonyms: Nuclear factor NF-kappa-B p65 subunit; NFKBP65; RELA; p65; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4149 Supplier Biomatik Corporation Supplier No. EKU08192
Human Transcription Factor P65 (RELA) ELISA Kit for detecting RELA in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04206. Gene ID: 5970. Target Synonyms: Nuclear factor NF-kappa-B p65 subunit; NFKBP65; RELA; p65; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.