Learn More
Biomatik Corporation Human V-Set Domain Containing T-Cell Activation Inhibitor 1 (VTCN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08196
Encompass_Preferred
Human V-Set Domain Containing T-Cell Activation Inhibitor 1 (VTCN1) ELISA Kit for detecting VTCN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7Z7D3. Target Synonyms: B7-H4; B7H4; B7S1; B7X; B7h.5; VCTN1; T-cell costimulatory molecule B7x; Immune costimulatory protein B7-H4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4150