Learn More
Biomatik Corporation Rat Complement Component 3 (C3) ELISA Kit
Supplier:  Biomatik Corporation EKU09030
Encompass_Preferred
Rat Complement Component 3 (C3) ELISA Kit for detecting C3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01026. Gene ID: 24232. Target Synonyms: ASP; CPAMD1; Complement Protein C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Acylation stimulating protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6567