Learn More
Biomatik Corporation Human WAP Four Disulfide Core Domain Protein 2 (WFDC2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08198
Encompass_Preferred
Human WAP Four Disulfide Core Domain Protein 2 (WFDC2) ELISA Kit for detecting WFDC2 in serum, plasma, tissue homogenates, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14508. Gene ID: 10406. Target Synonyms: WFDC2; HE4; WAP5; EDDM4; Epididymal Protein 4; Epididymal Secretory Protein E4; Putative Protease Inhibitor WAP5; Major epididymis-specific protein E4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4151