Learn More
Biomatik Corporation Dog Neutrophil gelatinase-associated lipocalin (NGAL) ELISA Kit
Supplier:  Biomatik Corporation EKU09062
Encompass_Preferred
Dog Neutrophil gelatinase-associated lipocalin (NGAL) ELISA Kit for detecting NGAL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.66ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A8I3Q9B0. Target Synonyms: Lipocalin-2; LCN2; p25; Lipocalin 2; Oncogene 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; Siderocalin LCN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6572