Learn More
Biomatik Corporation Human Wilms Tumor Protein (WT1) ELISA Kit, 96 tests
Catalog No. 501504152
Encompass_Preferred
Human Wilms Tumor Protein (WT1) ELISA Kit for detecting WT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19544. Gene ID: 7490. Target Synonyms: WAGR; GUD; WIT-2; WT33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4152 Supplier Biomatik Corporation Supplier No. EKU08200
Human Wilms Tumor Protein (WT1) ELISA Kit for detecting WT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19544. Gene ID: 7490. Target Synonyms: WAGR; GUD; WIT-2; WT33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.