Learn More
Biomatik Corporation Human Wingless Type MMTV Integration Site Family, Member 10B (WNT10B) ELISA Kit, 96 tests
Catalog No. 501504154
Encompass_Preferred
Human Wingless Type MMTV Integration Site Family, Member 10B (WNT10B) ELISA Kit for detecting WNT10B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00744. Gene ID: 7480. Target Synonyms: WNT-12; SHFM6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4154 Supplier Biomatik Corporation Supplier No. EKU08203
Human Wingless Type MMTV Integration Site Family, Member 10B (WNT10B) ELISA Kit for detecting WNT10B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00744. Gene ID: 7480. Target Synonyms: WNT-12; SHFM6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.