Learn More
Biomatik Corporation Human Wingless Type MMTV Integration Site Family, Member 11 (WNT11) ELISA Kit, 96 tests
Catalog No. 501504155
Encompass_Preferred
Human Wingless Type MMTV Integration Site Family, Member 11 (WNT11) ELISA Kit for detecting WNT11 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O96014. Gene ID: 7481. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4155 Supplier Biomatik Corporation Supplier No. EKU08205
Human Wingless Type MMTV Integration Site Family, Member 11 (WNT11) ELISA Kit for detecting WNT11 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O96014. Gene ID: 7481. Restrictions: For Research Use Only. Not for use in diagnostic procedures.