Learn More
Biomatik Corporation Mouse Guanylate Cyclase Activator 2A (GUCA2A) ELISA Kit
Catalog No. 502286593
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Mouse. Detection Range: 7.8-500pg/mL. Sensitivity: 3.1pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P33680. Target Synonyms: GUCA2; STARA; GCAP-I; Guanylin; Guanylate cyclase-activating protein 1. Restriction: For Research Use Only.
Catalog No. 50-228-6593 Supplier Biomatik Corporation Supplier No. EKU09229
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Mouse. Detection Range: 7.8-500pg/mL. Sensitivity: 3.1pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P33680. Target Synonyms: GUCA2; STARA; GCAP-I; Guanylin; Guanylate cyclase-activating protein 1. Restriction: For Research Use Only.