Learn More
Biomatik Corporation Rat NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) ELISA Kit
Supplier:  Biomatik Corporation EKU09287
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: AVP; AGTAVPRL; AII; AII/AVP; C1orf7; CIAS1; CLR1.1; FCAS; FCU; MWS; NALP3; PYPAF1; Cryopyrin; Cold Induced Autoinflammatory Syndrome 1; Caterpiller-Like Receptor 1.1. Restriction: For Research Use Only.
Catalog No. 50-228-6599