Learn More
Biomatik Corporation Human WNT1 Inducible Signaling Pathway Protein 2 (WISP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08226
Encompass_Preferred
Human WNT1 Inducible Signaling Pathway Protein 2 (WISP2) ELISA Kit for detecting WISP2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O76076. Gene ID: 8839. Target Synonyms: CCN5; CT58; CTGF-L; CCN family member 5; Connective tissue growth factor-like protein; Connective tissue growth factor-related protein 58. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4161