Learn More
Biomatik Corporation Human Xanthine Dehydrogenase (XDH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08232
Encompass_Preferred
Human Xanthine Dehydrogenase (XDH) ELISA Kit for detecting XDH in serum, plasma, breast milk, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47989. Gene ID: 7498. Target Synonyms: XO; XOR; XOD; Xanthine Oxidase; NAD+-Xanthine Dehydrogenase; Xanthine-NAD+ Oxidoreductase; Xanthine/NAD+ Oxidoreductase; Xanthine Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4163