Learn More
Biomatik Corporation Human Xeroderma Pigmentosum, Complementation Group C (XPC) ELISA Kit, 96 tests
Catalog No. 501504164
Encompass_Preferred
Human Xeroderma Pigmentosum, Complementation Group C (XPC) ELISA Kit for detecting XPC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01831. Gene ID: 7508. Target Synonyms: XP-C; XP3; XPCC; RAD4; Xeroderma Pigmentosum Group C Protein; DNA repair protein complementing XP-C cells. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4164 Supplier Biomatik Corporation Supplier No. EKU08238
Human Xeroderma Pigmentosum, Complementation Group C (XPC) ELISA Kit for detecting XPC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01831. Gene ID: 7508. Target Synonyms: XP-C; XP3; XPCC; RAD4; Xeroderma Pigmentosum Group C Protein; DNA repair protein complementing XP-C cells. Restrictions: For Research Use Only. Not for use in diagnostic procedures.