Learn More
Biomatik Corporation Human X-Ray Repair Cross Complementing 6 (XRCC6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08245
Encompass_Preferred
Human X-Ray Repair Cross Complementing 6 (XRCC6) ELISA Kit for detecting XRCC6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12956. Gene ID: 2547. Target Synonyms: Ku70; CTC75; CTCBF; G22P1; ML8; TLAA; Lupus Ku Autoantigen Protein P70; ATP-Dependent DNA Helicase 2 Subunit 1; CTC box-binding factor 75 kDa; Thyroid-lupus autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4166