Learn More
Biomatik Corporation High Sensitive Human Albumin (ALB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08258
Encompass_Preferred
The High Sensitive Human Albumin (ALB) ELISA Kit allows for quantitative determination of Albumin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02768. Cross Activity: No significant cross-reactivity or interference between Albumin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4170