Learn More
Biomatik Corporation Human/Mouse/Rat/Rabbit/Goat/Horse Brain Derived Neurotrophic Factor (BDNF) ELISA Kit
Supplier:  Biomatik Corporation EKU10071
Encompass_Preferred
Human/Mouse/Rat/Rabbit/Goat/Horse Brain Derived Neurotrophic Factor (BDNF) ELISA Kit for detecting BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Goat (Capra aegagrus hircus), Horse (Equus caballus; Equine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23560. Gene ID: 627. Target Synonyms: Neurotrophin; Abrineurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6648